missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.333 N (N/3), Ricca Chemical

$78.21 - $213.13

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40 g/mol
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7377-32
SDS View Documents
Ricca Chemical Company
737732
1 L Natural Poly Bottle
Each for $109.37
Only null left
 
7377-1
View Documents
Ricca Chemical Company
73771
4 L Natural Poly Bottle
Each for $213.13
Only null left
 
737716
SDS View Documents
Ricca Chemical Company
737716
500 mL Natural Poly Bottle
Each for $78.21
Only null left
 
Description

Description

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability
Specifications

Chemical Identifiers

1310-73-2
40 g/mol
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide

Specifications

1310-73-2
40 g/mol
0.333N (N/3)
Sodium Hydroxide
1 L
Laboratory
Natural Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'