missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.333 N (N/3), Ricca Chemical
Supplier: Ricca Chemical Company 73771
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Specifications
| Sodium Hydroxide | |
| 4 L | |
| Laboratory | |
| Natural Poly Bottle |
| 1310-73-2 | |
| 40 g/mol | |
| 0.333N (N/3) |
Chemical Identifiers
| 1310-73-2 | |
| 40 g/mol |