missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.313 Normal Suitable for Crude Fiber Analysis of Food and Feed, Reagents

Catalog No. 01336278
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336278 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336278 Supplier Labchem Inc Supplier No. C731824020F
Only null left

Sodium Hydroxide, 0.313 Normal Suitable for Crude Fiber Analysis of Food and Feed, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH >13
Color Colorless
Boiling Point 100°C
Quantity 20 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.014
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 0.313 Normal Suitable for Crude Fiber Analysis of Food and Feed