missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.3125 N, Ricca Chemical

Catalog No. 73745
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Poly Natural Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
73745 20 L Cubitainer
737432 1 L Poly Natural Bottle
73741 4 L Natural Poly Bottle
737425 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 73745 Supplier Ricca Chemical Company Supplier No. 73745
Only null left

Suitable for USDA Acidity Testing in Citrus Products

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40 g/mol
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Quantity 20 L
IUPAC Name sodium;hydroxide
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.3125N
Packaging Cubitainer
Chemical Name or Material Sodium Hydroxide, 0.3125N