missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.200 N (N/5), Ricca Chemical

Supplier:  Ricca Chemical Company 736016

 Afficher plus de versions de ce produit

Numéro de catalogue. 736016


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability
Spécifications

Spécifications

Sodium Hydroxide
1310-73-2,7732-18-5
100%,1.61
0.200Normal
500 mL
[OH-].[Na+]
40 g/mol
CHEBI:32145
Natural Poly Bottle
Colorless
96%,1.55
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Laboratory
Liquid

Identifiants chimiques

1310-73-2
40 g/mol
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS Documents

Documents

Promotions

Promotions