missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.200 N (N/5), Ricca Chemical

Catalog No. p-2917084
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
736032 1 L Natural Poly Bottle
73601 4 L Natural Poly Bottle
73605 20 L Cubitainer
736016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 736032 Supplier Ricca Chemical Company Supplier No. 736032
Only null left
  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 1310-73-2
Molecular Weight (g/mol) 40 g/mol

Specifications

Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.200N (N/5)
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide