missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.200 N (N/5), Ricca Chemical

Supplier:  Ricca Chemical Company 73601

 View more versions of this product

Catalog No. 7360-1

Only null left
Explore available promotions

Description

Description

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability
Specifications

Specifications

Sodium Hydroxide
4 L
Laboratory
Natural Poly Bottle
1310-73-2
40 g/mol
0.200N (N/5)

Chemical Identifiers

1310-73-2
40 g/mol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions