missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 0.1N Standardized Solution

Catalog No. AA35625K2
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35625K2 1 L
AA35625K7 4 L
AA35625M4 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35625K2 Supplier Thermo Scientific Chemicals Supplier No. 035625K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Sodium hydroxide, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1.1g/cm3 at 20°C
Physical Form Liquid
Solubility Miscible with glycerol, ethanol and methanol; Immiscible with acetone and ether
Type Analytical Volumetric Solution
Quantity 1 L
Formula Weight 40
Grade Standard
Concentration or Composition (by Analyte or Components) Sodium hydroxide: 0.4%; Water: 99.6%
Chemical Name or Material Sodium hydroxide
EINECSNumber 215-185-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only