missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.111 N (N/9), Ricca Chemical

Catalog No. 73541
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73541 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 73541 Supplier Ricca Chemical Company Supplier No. RI73541
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 98.0, 0.44
CAS Max % 100.0, 0.46
Grade Laboratory
Packaging Natural Poly Bottle
pH >13
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 0.111Normal
Chemical Name or Material Sodium Hydroxide