missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.100 N (N/10), Ricca Chemical
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Specifications
Specifications
| pH | 13 |
| Color | Colorless |
| Boiling Point | 100°C |
| Melting Point | 0°C |
| CAS Min % | 99.6, 0.4 |
| CAS Max % | 99.6, 0.4 |
| Density | 1.0046 |
| Solubility Information | Miscible in water |
| Quantity | 250 mL |
| Grade | Laboratory |
| Show More |
Safety and Handling
| ShelfLife | 24 months |
| Recommended Storage | Ambient |