missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.100 N (N/10), Ricca Chemical

Catalog No. R7350000120
Change view
Click to view available options
Quantity:
120 mL
250 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R7350000120 120 mL Natural Poly Bottle
R7350000250 250 mL Natural Poly Bottle
735016 500 mL Natural Poly Bottle
735032 1 L Natural Poly Bottle
73501CT 4 L Cubitainer
73501 4 L Natural Poly Bottle
735025 10 L Cubitainer
73505 20 L Cubitainer
RIC01002ST 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. R7350000120 Supplier Ricca Chemical Company Supplier No. R7350000120A
Only null left

Standardized Bases

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40 g/mol
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 13
Color Colorless
Boiling Point 100°C
Melting Point 0°C
CAS Min % 99.6, 0.4
CAS Max % 99.6, 0.4
Density 1.0046
Solubility Information Miscible in water
Quantity 120 mL
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient