missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.100 N (N/10), Ricca Chemical

Catalog No. 735032
Change view
Click to view available options
Quantity:
120 mL
250 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
735032 1 L Natural Poly Bottle
R7350000120 120 mL Natural Poly Bottle
R7350000250 250 mL Natural Poly Bottle
735016 500 mL Natural Poly Bottle
73501CT 4 L Cubitainer
73501 4 L Natural Poly Bottle
735025 10 L Cubitainer
73505 20 L Cubitainer
RIC01002ST 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 735032 Supplier Ricca Chemical Company Supplier No. 735032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Standardized Bases

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40 g/mol
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Min % 97%, 0.78
CAS Max % 100%, 0.81
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.100N (N/10)
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide Solution