missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.100 Normal (N/10), Reagents

Catalog No. 01336303
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
10 L
20 L
200 L
Packaging:
Cubitainer
Poly Bottle
Poly Drum
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336303 100 mL Poly Bottle
01336302 10 L Cubitainer
01336301 1 L Poly Bottle
01336300 200 L Poly Drum
01336299 20 L Cubitainer
01336298 4 L Poly Bottle
01336297 4 L Cubitainer
01336296 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01336303 Supplier Labchem Inc Supplier No. C7318000100A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Hydroxide, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH 13
Color Colorless
Boiling Point 100°C
Quantity 100 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0046
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 0.100 Normal (N/10)