missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.100 Normal (N/10) in Reagent Alcohol, Reagents

Catalog No. p-200021650
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333570 1 L
01333569 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333570 Supplier Labchem Inc Supplier No. CS1595001A
Only null left

Sodium Hydroxide, 0.100 Normal (N/10) in Reagent Alcohol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name ethanol
SMILES CCO

Specifications

pH 13
Color Colorless
Boiling Point 72°C
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.82
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 0.100 Normal (N/10) in Reagent Alcohol