missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 0.05N Standardized Solution

Catalog No. AA35623M4
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35623M4 20 L
AA35623K2 1 L
AA35623K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35623M4 Supplier Thermo Scientific Chemicals Supplier No. 035623M4
Only null left

CAS: 1310-73-2 | HNaO | 39.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Concentration 0.05N
Formula Weight 40
Odor Odorless
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Sodium hydroxide, Standardized Solution
EINECSNumber 215-185-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only