missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.0500 N (N/20), Ricca Chemical
$57.81 - $173.78
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 40 g/mol |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
733016
|
Ricca Chemical Company
733016 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
|
7330-32
|
Ricca Chemical Company
733032 |
1 L | Natural Poly Bottle |
N/A
|
|
||||
|
R73300004F
|
Ricca Chemical Company
R73300004F |
4 L | Cubitainer |
N/A
|
|
||||
|
73301
|
Ricca Chemical Company
73301 |
4 L | Natural Poly Bottle |
N/A
|
|
||||
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Chemical Identifiers
| 1310-73-2 | |
| 40 g/mol | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Specifications
| 1310-73-2,7732-18-5 | |
| 100%,0.41 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| Laboratory | |
| Natural Poly Bottle |
| 98%,0.39 | |
| HNaO | |
| 500 mL | |
| [OH-].[Na+] | |
| 40 g/mol | |
| CHEBI:32145 | |
| 0.0500N | |
| Sodium Hydroxide |