missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.0500 N (N/20), Ricca Chemical

Supplier:  Ricca Chemical Company 733032

 View more versions of this product

Catalog No. 7330-32


Only null left
Explore available promotions

Description

Description

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability
Specifications

Specifications

Sodium Hydroxide
98%,0.39
HNaO
1 L
[OH-].[Na+]
40 g/mol
CHEBI:32145
0.0500N
1310-73-2,7732-18-5
100%,0.41
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Laboratory
Natural Poly Bottle

Chemical Identifiers

1310-73-2
40 g/mol
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions