missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrosulfite, ca. 85%, Tech.

CAS: 7775-14-6 | Na2O4S2 | 174.1 g/mol

$59.68 - $313.96

Chemical Identifiers

CAS 7775-14-6
MDL Number MFCD00011640
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
PubChem CID 24489
ChEBI CHEBI:66870
SMILES [O-]S(=O)S(=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169590250
SDS View Documents
Thermo Scientific Chemicals
169590250
25 g Glass bottle
Each for $59.68
Only null left
 
AC169590010
SDS View Documents
Thermo Scientific Chemicals
169590010
1 kg Glass Bottle
Each for $190.96
Only null left
 
AC169590020
SDS View Documents
Thermo Scientific Chemicals
169590020
2 kg Glass Bottle
Each for $313.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

7775-14-6
JVBXVOWTABLYPX-UHFFFAOYSA-L
24489
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
MFCD00011640
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
CHEBI:66870

Specifications

>300.0°C
White
25 g
Na2O4S2
MFCD00011640
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
JVBXVOWTABLYPX-UHFFFAOYSA-L
174.1
CHEBI:66870
∼85%
Glass bottle
7775-14-6
Crystalline Powder
ca. 85%
Na2S2O4
15, 8747
Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
24489
174.1
Technical
Sodium hydrosulfite, tech., ca. 85%
Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Self-heating; may catch fire. Contact with acids liberates toxic gas.

GHS P Statement Keep cool. Protect from sunlight. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger

EINECSNumber : 231-890-0

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only