Learn More
Sodium hydrosulfite, ca. 85%, Tech.
CAS: 7775-14-6 | Na2O4S2 | 174.1 g/mol
Supplier: Thermo Scientific Chemicals 169590020
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
| Sodium hydrosulfite | |
| tech., ca. 85% | |
| White | |
| 2 kg | |
| Na2O4S2 | |
| MFCD00011640 | |
| sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros | |
| JVBXVOWTABLYPX-UHFFFAOYSA-L | |
| 174.096 | |
| CHEBI:66870 | |
| ∼85% | |
| Glass Bottle |
| >300.0°C | |
| 7775-14-6 | |
| Crystalline Powder | |
| ca. 85% | |
| Na2S2O4 | |
| 15, 8747 | |
| Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol | |
| [O-]S(=O)S(=O)[O-].[Na+].[Na+] | |
| 24489 | |
| 174.1 | |
| Technical |
Chemical Identifiers
| 7775-14-6 | |
| JVBXVOWTABLYPX-UHFFFAOYSA-L | |
| 24489 | |
| [O-]S(=O)S(=O)[O-].[Na+].[Na+] |
| MFCD00011640 | |
| sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros | |
| CHEBI:66870 |
Safety and Handling
GHS P Statement Harmful if swallowed. Self-heating; may catch fire. Contact with acids liberates toxic gas.
GHS P Statement Keep cool. Protect from sunlight. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Danger
EINECSNumber : 231-890-0
TSCA : TSCA
RUO – Research Use Only