missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrosulfite, ca. 85%, Tech.

CAS: 7775-14-6 | Na2O4S2 | 174.1 g/mol

Supplier:  Thermo Scientific Chemicals 169590010

 View more versions of this product

Catalog No. AC169590010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Sodium hydrosulfite
tech., ca. 85%
White
1 kg
Na2O4S2
MFCD00011640
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
JVBXVOWTABLYPX-UHFFFAOYSA-L
174.096
CHEBI:66870
∼85%
Glass Bottle
>300.0°C
7775-14-6
Crystalline Powder
ca. 85%
Na2S2O4
15, 8747
Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
24489
174.1
Technical

Chemical Identifiers

7775-14-6
JVBXVOWTABLYPX-UHFFFAOYSA-L
24489
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
MFCD00011640
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
CHEBI:66870
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Self-heating; may catch fire. Contact with acids liberates toxic gas.

GHS P Statement Keep cool. Protect from sunlight. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger

EINECSNumber : 231-890-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only