missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrogen Carbonate/Sodium Carbonate, For pH Measurement, Certipur™, MilliporeSigma™
Specifications
Specifications
| Color | White powder |
| Formula Weight | 84.01 |
| Odor | Odorless |
| Packaging | Fibre case |
| Quantity | 2 x 25 g |
| Physical Form | Powder |
| Chemical Name or Material | Sodium Hydrogen Carbonate/Sodium Carbonate, Certified secondary standard reference material for pH measurement; directly traceable to primary SRM from NIST/PTB pH(S) = 10.012 (25°C). |