missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen arsenate heptahydrate, 98%

CAS: 10048-95-0 | AsH15Na2O11 | 312.01 g/mol

$159.01 - $563.70

Chemical Identifiers

CAS 10048-95-0
Molecular Formula AsH15Na2O11
Molecular Weight (g/mol) 312.01
MDL Number MFCD00149179
InChI Key KOLXPEJIBITWIQ-UHFFFAOYSA-L
Synonym disodium arsenate heptahydrate, sodium acid arsenate, heptahydrate, sodium arsenate heptahydrate, sodium orthoarsenate heptahydrate, dibasic sodium arsenate heptahydrate, sodium arsenate, dibasic, heptahydrate, unii-90zq7g407w, disodium arsenate, heptahydrate, ccris 5553, arsenic acid h3aso4 , disodium salt, heptahydrate
PubChem CID 61460
IUPAC Name disodium;hydrogen arsorate;heptahydrate
SMILES O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1827522
View Documents
Thermo Scientific Chemicals
A1827522
100 g
Each for $159.01
Only null left
 
AAA1827536
View Documents
Thermo Scientific Chemicals
A1827536
500 g
Each for $563.70
Only null left
 
Description

Description

Sodium hydrogen arsenate heptahydrate is used as a source of soluble arsenic and in dyeing and printing with Turkey-red oil. It also acts as a veterinary drug for anti-parasitic, skin diseases, blood diseases, rheumatism, and asthma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and glycerol.

Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications

Chemical Identifiers

10048-95-0
312.01
KOLXPEJIBITWIQ-UHFFFAOYSA-L
61460
O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O
AsH15Na2O11
MFCD00149179
disodium arsenate heptahydrate, sodium acid arsenate, heptahydrate, sodium arsenate heptahydrate, sodium orthoarsenate heptahydrate, dibasic sodium arsenate heptahydrate, sodium arsenate, dibasic, heptahydrate, unii-90zq7g407w, disodium arsenate, heptahydrate, ccris 5553, arsenic acid h3aso4 , disodium salt, heptahydrate
disodium;hydrogen arsorate;heptahydrate

Specifications

130°C (decomposition)
100 g
MFCD00149179
disodium arsenate heptahydrate, sodium acid arsenate, heptahydrate, sodium arsenate heptahydrate, sodium orthoarsenate heptahydrate, dibasic sodium arsenate heptahydrate, sodium arsenate, dibasic, heptahydrate, unii-90zq7g407w, disodium arsenate, heptahydrate, ccris 5553, arsenic acid h3aso4 , disodium salt, heptahydrate
KOLXPEJIBITWIQ-UHFFFAOYSA-L
disodium;hydrogen arsorate;heptahydrate
61460
98%
1.88
10048-95-0
AsH15Na2O11
UN1685
Soluble in water and glycerol.
O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O
312.01
312.01 (185.94 Anhydrous)
Odorless
Sodium hydrogen arsenate heptahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SODIUM ARSENATE

EINECSNumber : 231-902-4

RTECSNumber : CG0900000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only