Learn More
Sodium hydrogen arsenate heptahydrate, 98%
CAS: 10048-95-0 | AsH15Na2O11 | 312.01 g/mol
Supplier: Thermo Scientific Chemicals A1827522
Description
Sodium hydrogen arsenate heptahydrate is used as a source of soluble arsenic and in dyeing and printing with Turkey-red oil. It also acts as a veterinary drug for anti-parasitic, skin diseases, blood diseases, rheumatism, and asthma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water and glycerol.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
| Sodium hydrogen arsenate heptahydrate | |
| 10048-95-0 | |
| AsH15Na2O11 | |
| UN1685 | |
| Soluble in water and glycerol. | |
| O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O | |
| 312.01 | |
| 312.01 (185.94 Anhydrous) | |
| Odorless |
| 130°C (decomposition) | |
| 100 g | |
| MFCD00149179 | |
| disodium arsenate heptahydrate, sodium acid arsenate, heptahydrate, sodium arsenate heptahydrate, sodium orthoarsenate heptahydrate, dibasic sodium arsenate heptahydrate, sodium arsenate, dibasic, heptahydrate, unii-90zq7g407w, disodium arsenate, heptahydrate, ccris 5553, arsenic acid h3aso4 , disodium salt, heptahydrate | |
| KOLXPEJIBITWIQ-UHFFFAOYSA-L | |
| disodium;hydrogen arsorate;heptahydrate | |
| 61460 | |
| 98% | |
| 1.88 |
Chemical Identifiers
| 10048-95-0 | |
| 312.01 | |
| KOLXPEJIBITWIQ-UHFFFAOYSA-L | |
| 61460 | |
| O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O |
| AsH15Na2O11 | |
| MFCD00149179 | |
| disodium arsenate heptahydrate, sodium acid arsenate, heptahydrate, sodium arsenate heptahydrate, sodium orthoarsenate heptahydrate, dibasic sodium arsenate heptahydrate, sodium arsenate, dibasic, heptahydrate, unii-90zq7g407w, disodium arsenate, heptahydrate, ccris 5553, arsenic acid h3aso4 , disodium salt, heptahydrate | |
| disodium;hydrogen arsorate;heptahydrate |
Safety and Handling
GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SODIUM ARSENATE
EINECSNumber : 231-902-4
RTECSNumber : CG0900000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only