accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OH2ONaNaOHAsOOO

CAS RN 10048-95-0

IUPAC Name: disodium heptahydrate hydrogen arsorate
Synonyms: disodium heptahydrate hydrogen arsenate
Molecular Weight (g/mol): 312.01
Molecular Formula: AsH15Na2O11
InChi Key: KOLXPEJIBITWIQ-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O
Grade 
  • (2)
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (6)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (6)
  • (4)
  • (2)
  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)