missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydride, 60% Dispersion in Mineral Oil, Spectrum™ Chemical

S1288, 7646-69-7, NaH

Supplier:  Spectrum Chemical Mfg Cor S1288100GM

 View more versions of this product

Catalog No. 18606519

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Hydride, 60 Percent Dispersion in Mineral Oil , a chemical compound, can be used in organic synthesis as a strong base, as well as a drying agent to dry organic solvents. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Specifications

7646-69-7
100 g
UN1427
[Na]
22.99
1
Na
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
Reagent

Chemical Identifiers

7646-69-7
22.99
sodium
Na
KEAYESYHFKHZAL-UHFFFAOYSA-N
[Na]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions