missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydride, 60% Dispersion in Mineral Oil, Spectrum™ Chemical

Catalog No. p-7158617
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
12 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18606519 100 g
18606520 500 g
18606521 1 kg
18606522 12 kg
18606523 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606519 Supplier Spectrum Chemical Mfg Cor Supplier No. S1288100GM
Only null left

S1288, 7646-69-7, NaH

Spectrum™ Chemical Sodium Hydride, 60 Percent Dispersion in Mineral Oil , a chemical compound, can be used in organic synthesis as a strong base, as well as a drying agent to dry organic solvents. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7646-69-7
Molecular Formula Na
Molecular Weight (g/mol) 22.99
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
IUPAC Name sodium
SMILES [Na]

Specifications

CAS Max % 1
Quantity 100 g
UN Number UN1427
Grade Reagent