accessibility menu, dialog, popup

UserName

Na

CAS RN 7646-69-7

IUPAC Name: sodium
Synonyms: sodium sodio sodium atom natrium
Molecular Weight (g/mol): 22.99
Molecular Formula: Na
InChi Key: KEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES: [Na]
Grade 
  • (1)
  • (7)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Physical Form 
  • (5)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (7)
  • (5)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)