missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hippurate 98.0+%, TCI America™
Supplier: TCI America H0144500G
Specifications
| Sodium Hippurate | |
| White | |
| MFCD00002693 | |
| sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt | |
| C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+] | |
| 201.157 | |
| 201.16 | |
| Crystalline Powder |
| 532-94-5 | |
| C9H8NNaO3 | |
| 500 g | |
| ZBCAZEFVTIBZJS-UHFFFAOYSA-M | |
| sodium;2-benzamidoacetate | |
| 516953 | |
| ≥98.0% (T) |
Chemical Identifiers
| 532-94-5 | |
| 201.157 | |
| ZBCAZEFVTIBZJS-UHFFFAOYSA-M | |
| 516953 | |
| C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+] |
| C9H8NNaO3 | |
| MFCD00002693 | |
| sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt | |
| sodium;2-benzamidoacetate |
Safety and Handling
RTECSNumber : MR8170000
TSCA : Yes