missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hippurate 98.0+%, TCI America™

Catalog No. p-7130370
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H014425G 25 g
H0144500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H014425G Supplier TCI America Supplier No. H014425G
Only null left

Chemical Identifiers

CAS 532-94-5
Molecular Formula C9H8NNaO3
Molecular Weight (g/mol) 201.157
MDL Number MFCD00002693
InChI Key ZBCAZEFVTIBZJS-UHFFFAOYSA-M
Synonym sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt
PubChem CID 516953
IUPAC Name sodium;2-benzamidoacetate
SMILES C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 201.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Hippurate
RTECSNumber MR8170000
TSCA Yes