missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethanesulfonate 98.0+%, TCI America™

Supplier:  TCI America E025625G

 View more versions of this product

Catalog No. E025625G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Ethanesulfonate
270°C
C2H5NaO3S
25 g
KQFAFFYKLIBKDE-UHFFFAOYSA-M
sodium;ethanesulfonate
4192170
≥98.0% (T)
5324-47-0
White
MFCD00066498
sodium ethanesulfonate, sodium ethanesulphonate, ethanesulfonic acid sodium salt, ethanesulfonic acid, sodium salt, ethane sulfonic acid sodium salt, sodiumethanesulfonate, sodium ethylsulfonate, sodium esylate, sodium ethane sulfonate, ethylsulfonic acid sodium salt
CCS(=O)(=O)[O-].[Na+]
132.109
132.11
Crystalline Powder

Chemical Identifiers

5324-47-0
132.109
KQFAFFYKLIBKDE-UHFFFAOYSA-M
4192170
CCS(=O)(=O)[O-].[Na+]
C2H5NaO3S
MFCD00066498
sodium ethanesulfonate, sodium ethanesulphonate, ethanesulfonic acid sodium salt, ethanesulfonic acid, sodium salt, ethane sulfonic acid sodium salt, sodiumethanesulfonate, sodium ethylsulfonate, sodium esylate, sodium ethane sulfonate, ethylsulfonic acid sodium salt
sodium;ethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions