missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethanesulfonate 98.0+%, TCI America™

$167.76 - $419.41

Chemical Identifiers

CAS 5324-47-0
Molecular Formula C2H5NaO3S
Molecular Weight (g/mol) 132.109
MDL Number MFCD00066498
InChI Key KQFAFFYKLIBKDE-UHFFFAOYSA-M
Synonym sodium ethanesulfonate, sodium ethanesulphonate, ethanesulfonic acid sodium salt, ethanesulfonic acid, sodium salt, ethane sulfonic acid sodium salt, sodiumethanesulfonate, sodium ethylsulfonate, sodium esylate, sodium ethane sulfonate, ethylsulfonic acid sodium salt
PubChem CID 4192170
IUPAC Name sodium;ethanesulfonate
SMILES CCS(=O)(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E02565G
View Documents
TCI America
E02565G
5 g
Each for $167.76
Only null left
 
E025625G
View Documents
TCI America
E025625G
25 g
Each for $419.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5324-47-0
132.109
KQFAFFYKLIBKDE-UHFFFAOYSA-M
4192170
CCS(=O)(=O)[O-].[Na+]
C2H5NaO3S
MFCD00066498
sodium ethanesulfonate, sodium ethanesulphonate, ethanesulfonic acid sodium salt, ethanesulfonic acid, sodium salt, ethane sulfonic acid sodium salt, sodiumethanesulfonate, sodium ethylsulfonate, sodium esylate, sodium ethane sulfonate, ethylsulfonic acid sodium salt
sodium;ethanesulfonate

Specifications

5324-47-0
White
MFCD00066498
sodium ethanesulfonate, sodium ethanesulphonate, ethanesulfonic acid sodium salt, ethanesulfonic acid, sodium salt, ethane sulfonic acid sodium salt, sodiumethanesulfonate, sodium ethylsulfonate, sodium esylate, sodium ethane sulfonate, ethylsulfonic acid sodium salt
CCS(=O)(=O)[O-].[Na+]
132.109
132.11
Crystalline Powder
270°C
C2H5NaO3S
5 g
KQFAFFYKLIBKDE-UHFFFAOYSA-M
sodium;ethanesulfonate
4192170
≥98.0% (T)
Sodium Ethanesulfonate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'