missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Dodecyl Sulfate, OmniPur™, Calbiochem™,
$395.87
Chemical Identifiers
| CAS | 151-21-3 |
|---|---|
| Molecular Formula | C12H25NaO4S |
| Molecular Weight (g/mol) | 288.38 |
| MDL Number | MFCD00036175 |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| Synonym | SDS, Sodium Lauryl Sulfate, n-Dodecyl Sulfate, Sodium |
| PubChem CID | 3423265 |
| ChEBI | CHEBI:8984 |
| IUPAC Name | sodium dodecyl sulfate |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Chemical Identifiers
| C12H25NaO4S | |
| MFCD00036175 | |
| SDS, Sodium Lauryl Sulfate, n-Dodecyl Sulfate, Sodium | |
| CHEBI:8984 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Specifications
| White | |
| Fine powder | |
| MFCD00036175 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 3423265 | |
| ≥99.0% (dried basis) | |
| Sodium Dodecyl Sulfate |
| 151-21-3 | |
| C12H25NaO4S | |
| SDS, Sodium Lauryl Sulfate, n-Dodecyl Sulfate, Sodium | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| CHEBI:8984 | |
| Molecular Biology |
Safety and Handling
storageNote1 : Ok to freeze