missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Sodium Dodecyl Sulfate, OmniPur™, Calbiochem™,

Manufacturer:  MilliporeSigma™ 7910500GM

 View more versions of this product

Catalog No. 7910500GM



Description

Description

Recommended Storage

Ok to freeze

Specifications

Specifications

Sodium Dodecyl Sulfate
C12H25NaO4S
Plastic Bottle
CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
288.378
CHEBI:8984
≥99.0% (dried basis)
White
151-21-3
SDS, Sodium Lauryl Sulfate, n-Dodecyl Sulfate, Sodium
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium;dodecyl sulfate
3423265
Fine powder
Molecular Biology
500g

Chemical Identifiers

151-21-3
288.378
SDS, Sodium Lauryl Sulfate, n-Dodecyl Sulfate, Sodium
CHEBI:8984
CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
C12H25NaO4S
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
sodium;dodecyl sulfate
Safety and Handling

Safety and Handling

storageNote1 : Ok to freeze

SDS
Documents