missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate, Molecular Grade, Promega™ DFS Item

A detergent to denature proteins.

$80.37 - $292.51

Chemical Identifiers

CAS 7732-18-5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR-V6551 DFS Item
View Documents
Promega Corporation
V6551
100 mL
Each for $80.37
Only null left
 
PR-H5113 DFS Item
View Documents
Promega Corporation
H5113
100 g
Each for $110.68
Only null left
 
PRH5114 DFS Item
SDS View Documents
Promega Corporation
H5114
500 g
Each for $292.51
Only null left
 
Description

Description

  • Quality tested and certified free of DNase and RNase activity
Specifications

Chemical Identifiers

7732-18-5

Specifications

7732-18-5
100 mL
Molecular Biology
Sodium Dodecyl Sulfate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15 to 30°C

SDS
missing translation for 'documents'

missing translation for 'documents'