missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate, Molecular Grade, Promega™ DFS Item

A detergent to denature proteins.

Supplier:  Promega Corporation H5114

 View more versions of this product

Catalog No. PRH5114


Only null left
Explore available promotions

Description

Description

  • Quality tested and certified free of DNase and RNase activity
Specifications

Specifications

Sodium Dodecyl Sulfate
7732-18-5
500 g
Molecular Biology

Chemical Identifiers

7732-18-5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15 to 30°C

SDS Documents

Documents

Promotions

Promotions