missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dodecyl sulfate, >99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210291880
Specifications
| Sodium dodecyl sulfate | |
| White | |
| C12H25NaO4S | |
| 100 g | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium;dodecyl sulfate | |
| 3423265 | |
| 288.38 | |
| Granular Powder |
| 151-21-3 | |
| >1.1 at 20°C | |
| MFCD00036175 | |
| sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| CHEBI:8984 | |
| >99% |
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| 3423265 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| C12H25NaO4S | |
| MFCD00036175 | |
| sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
| CHEBI:8984 |