missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dodecyl sulfate, >99%, MP Biomedicals™
$69.85 - $751.83
Chemical Identifiers
| CAS | 151-21-3 |
|---|---|
| Molecular Formula | C12H25NaO4S |
| Molecular Weight (g/mol) | 288.38 |
| MDL Number | MFCD00036175 |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| Synonym | sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal |
| PubChem CID | 3423265 |
| ChEBI | CHEBI:8984 |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10291810
|
MP Biomedicals Inc
0210291810 |
10 g |
Each for $69.85
|
|
|||||
|
ICN10291880
|
MP Biomedicals Inc
0210291880 |
100 g |
Each for $140.51
|
|
|||||
|
ICN10291890
|
MP Biomedicals Inc
0210291890 |
500 g |
Each for $508.38
|
|
|||||
|
ICN10291891
|
MP Biomedicals Inc
0210291891 |
1 kg |
Each for $751.83
|
|
|||||
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| 3423265 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| C12H25NaO4S | |
| MFCD00036175 | |
| sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
| CHEBI:8984 |
Specifications
| 151-21-3 | |
| >1.1 at 20°C | |
| MFCD00036175 | |
| sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| CHEBI:8984 | |
| >99% | |
| Sodium dodecyl sulfate |
| White | |
| C12H25NaO4S | |
| 10 g | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 3423265 | |
| 288.38 | |
| Granular Powder |