missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, >99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210291890

 View more versions of this product

Catalog No. ICN10291890


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium dodecyl sulfate
White
C12H25NaO4S
500 g
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
288.38
Granular Powder
151-21-3
>1.1 at 20°C
MFCD00036175
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
>99%

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
C12H25NaO4S
MFCD00036175
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
CHEBI:8984
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions