missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dodecyl sulfate, 95%, MP Biomedicals™
Approximately 95% alkyl sulfate, predominantly composed of C12 and C14 homologs
$62.83 - $180.76
Chemical Identifiers
| CAS | 151-21-3 |
|---|---|
| Molecular Formula | C12H25NaO4S |
| Molecular Weight (g/mol) | 288.38 |
| MDL Number | MFCD00036175 |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| Synonym | SDS, Lauryl sulfate sodium salt |
| PubChem CID | 3423265 |
| ChEBI | CHEBI:8984 |
| IUPAC Name | sodium dodecyl sulfate |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19052280
|
MP Biomedicals Inc
0219052280 |
100 g |
Each for $62.83
|
|
|||||
|
ICN19052290
|
MP Biomedicals Inc
0219052290 |
500 g |
Each for $124.43
|
|
|||||
|
ICN19052291
|
MP Biomedicals Inc
0219052291 |
1 kg |
Each for $180.76
|
|
|||||
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| 3423265 | |
| sodium dodecyl sulfate |
| C12H25NaO4S | |
| MFCD00036175 | |
| SDS, Lauryl sulfate sodium salt | |
| CHEBI:8984 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Specifications
| White | |
| Granular Powder | |
| 100 g | |
| C12H25NaO4S | |
| SDS, Lauryl sulfate sodium salt | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| CHEBI:8984 | |
| Sodium dodecyl sulfate |
| 151-21-3 | |
| >1.1 at 20°C (water = 1) | |
| 0.95 | |
| MFCD00036175 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 3423265 | |
| 288.38 |