missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, 95%, MP Biomedicals™

Approximately 95% alkyl sulfate, predominantly composed of C12 and C14 homologs

Supplier:  MP Biomedicals Inc 0219052280

 View more versions of this product

Catalog No. ICN19052280


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium dodecyl sulfate
151-21-3
>1.1 at 20°C (water = 1)
0.95
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
288.38
White
Granular Powder
100 g
C12H25NaO4S
SDS, Lauryl sulfate sodium salt
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
sodium dodecyl sulfate
C12H25NaO4S
MFCD00036175
SDS, Lauryl sulfate sodium salt
CHEBI:8984
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions