missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dimethyl 5-Sulfoisophthalate 98.0+%, TCI America™

Catalog No. S015725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S015725G 25 g
S0157500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S015725G Supplier TCI America Supplier No. S015725G

Chemical Identifiers

CAS 3965-55-7
Molecular Formula C10H9NaO7S
Molecular Weight (g/mol) 296.225
MDL Number MFCD00007493
InChI Key LLHSEQCZSNZLRI-UHFFFAOYSA-M
Synonym sodium 3,5-bis methoxycarbonyl benzenesulfonate, sodium dimethyl 5-sulphonatoisophthalate, dimethyl 5-sulfoisophthalate sodium salt, unii-7l3g5x9vdu, sodium 1,3-dimethyl 5-sulfoisophthalate, 7l3g5x9vdu, 5-sulfoisophthalic acid, dimethyl ester, sodium salt, sodium dimethyl 5-sulfoisophthalate, 1,3-benzenedicarboxylic acid, 5-sulfo-, 1,3-dimethyl ester, sodium salt, 3,5-bis-methylkarboxy-benzensulfonan sodny czech
PubChem CID 23675796
IUPAC Name sodium;3,5-bis(methoxycarbonyl)benzenesulfonate
SMILES COC(=O)C1=CC(=CC(=C1)S(=O)(=O)[O-])C(=O)OC.[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 296.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dimethyl 5-Sulfoisophthalate
EINECSNumber (3)-1885&(3)-2134
RTECSNumber DB5044550
TSCA Yes