accessibility menu, dialog, popup

UserName

CAS RN 3965-55-7

NaCH3OOSOOOOOCH3

CAS RN 3965-55-7

IUPAC Name: sodium 3,5-bis(methoxycarbonyl)benzene-1-sulfonate
Synonyms: sodium 3,5-bis(methoxycarbonyl)benzenesulfonate
Molecular Weight (g/mol): 296.22
Molecular Formula: C10H9NaO7S
InChi Key: LLHSEQCZSNZLRI-UHFFFAOYSA-M
SMILES: [Na+].COC(=O)C1=CC(=CC(=C1)C(=O)OC)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results