missing translation for 'onlineSavingsMsg'
Learn More

Sodium Deoxycholate, 98%, Spectrum™ Chemical

SO166, 302-95-4, C24H39NaO4

$7396.54 - $14756.36

Chemical Identifiers

CAS 302-95-4
Molecular Formula C24H39NaO4
Molecular Weight (g/mol) 414.56
MDL Number MFCD00064139
InChI Key FHHPUSMSKHSNKW-SMOYURAASA-M
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606483
View Documents
Spectrum Chemical Mfg Cor
SO166500GM
500 g Poly Bottle
Each for $7,396.54
Only null left
 
18606484
View Documents
Spectrum Chemical Mfg Cor
SO1661KGBL
1 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Sodium Deoxycholate is a bile acid and a metabolic byproduct of intestinal bacteria. It is soluble in acetic acid and alcohol and appears as a white or off white crystalline powder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Chemical Identifiers

302-95-4
414.56
FHHPUSMSKHSNKW-SMOYURAASA-M
[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
C24H39NaO4
MFCD00064139
sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate

Specifications

302-95-4
500 g
MFCD00064139
[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
414.56
Reagent
1
C24H39NaO4
FHHPUSMSKHSNKW-SMOYURAASA-M
sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
98%
Poly Bottle
Videos
SDS
Documents

Documents