missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Deoxycholate, 98%, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Deoxycholate is a bile acid and a metabolic byproduct of intestinal bacteria. It is soluble in acetic acid and alcohol and appears as a white or off white crystalline powder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Spécifications
Spécifications
| CAS Max % | 1 |
| Quantity | 1 kg |
| Percent Purity | 98% |
| Grade | Reagent |
| Packaging | Poly Pail |