missing translation for 'onlineSavingsMsg'
Learn More

Sodium Deoxycholate, 98%, Spectrum™ Chemical

Catalog No. 18606483
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606483 500 g Poly Bottle
18606484 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606483 Supplier Spectrum Chemical Mfg Cor Supplier No. SO166500GM
Only null left

SO166, 302-95-4, C24H39NaO4

Spectrum™ Chemical Sodium Deoxycholate is a bile acid and a metabolic byproduct of intestinal bacteria. It is soluble in acetic acid and alcohol and appears as a white or off white crystalline powder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 302-95-4
Molecular Formula C24H39NaO4
Molecular Weight (g/mol) 414.56
MDL Number MFCD00064139
InChI Key FHHPUSMSKHSNKW-SMOYURAASA-M
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle