missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dehydroacetate, 98-100.5%, Spectrum™ Chemical
S1265, 64039-28-7, C8H7NaO4·H2O
$255.43 - $18035.97
Chemical Identifiers
| CAS | 64039-28-7 |
|---|---|
| Molecular Formula | C8H9NaO5 |
| Molecular Weight (g/mol) | 208.15 |
| InChI Key | AFIQQQWSAQPUQO-UHFFFAOYSA-N |
| IUPAC Name | sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide hydrate |
| SMILES | O.[Na+].CC(=O)[C-]1C(=O)OC(C)=CC1=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606477
|
Spectrum Chemical Mfg Cor
S1265125GM |
125 g | Poly Bottle |
Each for $255.43
|
|
||||
|
18606478
|
Spectrum Chemical Mfg Cor
S1265500GM |
500 g | Poly Bottle |
Each for $833.43
|
|
||||
|
18606479
|
Spectrum Chemical Mfg Cor
S126525KG |
2.5 kg | Poly Bottle |
Each for $3,086.46
|
|
||||
|
18606480
|
Spectrum Chemical Mfg Cor
S126550KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sodium DehydroacetateChemical Identifiers
| 64039-28-7 | |
| 208.15 | |
| sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide hydrate |
| C8H9NaO5 | |
| AFIQQQWSAQPUQO-UHFFFAOYSA-N | |
| O.[Na+].CC(=O)[C-]1C(=O)OC(C)=CC1=O |
Specifications
| 64039-28-7 | |
| Poly Bottle | |
| 125 g | |
| O.[Na+].CC(=O)[C-]1C(=O)OC(C)=CC1=O | |
| 208.15 | |
| Reagent |
| 1 | |
| C8H9NaO5 | |
| AFIQQQWSAQPUQO-UHFFFAOYSA-N | |
| sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide hydrate | |
| 98.0 to 100.5% |