missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dehydroacetate, 98-100.5%, Spectrum™ Chemical

Catalog No. 18606479
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606479 2.5 kg Poly Bottle
18606477 125 g Poly Bottle
18606478 500 g Poly Bottle
18606480 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606479 Supplier Spectrum Chemical Mfg Cor Supplier No. S12652.5KG
Only null left

S1265, 64039-28-7, C8H7NaO4·H2O

Spectrum™ Chemical Sodium Dehydroacetate

Chemical Identifiers

CAS 64039-28-7
Molecular Formula C8H9NaO5
Molecular Weight (g/mol) 208.15
InChI Key AFIQQQWSAQPUQO-UHFFFAOYSA-N
IUPAC Name sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide hydrate
SMILES O.[Na+].CC(=O)[C-]1C(=O)OC(C)=CC1=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 98.0 to 100.5%
Grade Reagent