missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, For Calcium, Magnesium, 2.5% (w/v), LabChem™

Catalog No. lc237007
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC237007 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC237007 Supplier LabChem Supplier No. LC237007
Only null left

Chemical Identifiers

CAS 7732-18-5 , 143-33-9 , 1310-73-2
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Color Colorless
CAS Max % 97.1, 2.5, 0.4
Linear Formula NaCN
UN Number UN3414
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 49
Percent Purity 2.5% (w/v)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Hydrogen cyanide; Nitrogen oxides
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Sodium Cyanide, For Calcium, Magnesium
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic if swallowed or in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid breathing mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Immediately call a poison center or doctor.
If on skin: Wash with plenty of soap and water.
Remove/Take off immediately all contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature