missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, 3.8% (w/v), Ricca Chemical

Catalog No. p-3252318
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
72284 120 mL Natural Poly Bottle
722816 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 72284 Supplier Ricca Chemical Company Supplier No. 72284
Only null left

Can be used as a whole blood sample anticoagulant solution use 0.5 mL per 4.5 mL whole blood. (Non-Regulated)

Chemical Identifiers

CAS 6132-04-3 , 7732-18-5
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

CAS Min % 94.0, 3.59
CAS Max % 98.0, 3.74
Percent Purity 3.8%
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼8
Quantity 120 mL
Chemical Name or Material Sodium Citrate (3.8% w/v)