missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chromate, 2% w/v, Reagents
Sodium Chromate, 2% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$131.36 - $290.04
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Chromium disodium oxide |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-333-635
|
Labchem Inc
CS14490010C |
10 mL |
Each for $131.36
|
|
|||||
|
01-333-634
|
Labchem Inc
CS1449001A |
1 L |
Each for $290.04
|
|
|||||
|
01-333-633
|
Labchem Inc
CS144900500A |
500 mL |
Each for $214.11
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Chromium disodium oxide | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| 100°C | |
| 7732-18-5,10034-82-9 | |
| 10 mL | |
| Chromium disodium oxide | |
| O | |
| 18.02 | |
| Reagent | |
| 1.02 |
| Yellow | |
| Liquid | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 2% | |
| Glass Bottle | |
| Sodium Chromate, 2% w/v |